Learn More
Enzo Life SciencesSupplier Diversity Partner Jurkat (heat shocked) , (cell lysate) (200 µg)  DFS Item
Manufacturer:  Enzo Life SciencesSupplier Diversity Partner ADILYCJK101F
Formulation: Liquid. In SDS-PAGE sample buffer, pH 6.8, heated for 10 min. at 70°C and containing 40mM TRIS-HCl, 1% SDS, 50mM DTT, 7.5% glycerol, and 0.003% bromophenol blue. Applications: WB. Shipping: Shipped on Dry Ice. Long Term Storage: -80°C. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-666-698