Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Justrite™ Sure-Grip™ EX Compac Corrosives/Acid Steel Safety Cabinet Available on GSA/VA Contract for Federal Government customers only.

Get more space to store hazardous material in your work environment. Justrite Sure-Grip™ EX Compac Corrosives/Acid Steel Safety Cabinet smaller sized cabinets fit nearly anywhere.

Manufacturer:  Justrite™ 8912022

 View more versions of this product

Catalog No. 19-159-719


  Check Availability
Add to cart

Alert:

These cabinets are not recommended for storage of phenol, nitric acid or sulfuric acid.


Specifications

Description

  • Flame-coated thermoplastic liner
  • 100% seamless liner on all interior walls, ceiling, sump, inside doors and shelves
  • Seam-free surface
  • Non-porous surface resists staining and cleans-up easily
  • Dual-benefits/Multi-use - complete protection from harsh acids and corrosive chemicals
  • Blue color

  • Features double-wall, 18-gauge (1mm) welded steel construction, 2 in. (51mm) liquid-tight containment sump, dual vents
  • Adjustable shelves, three-point stainless steel bullet self-latching doors with ULoc™ padlockable handle and Haz-Alert™ reflective warning labels
  • ChemCor™ liner interior

Compliance

Compliance: Retains FM Approval & Complies with OSHA regulations and the International Fire Code of NFPA 1 Uniform Fire Code. Only NFPA/OSHA for 19159712/19413871.

Description & Specifications

Specifications

12 gal., 45L
12 gal.
30.5 in.
CR Steel
50.8cm
Blue
14.56 in.
37cm
Flammables
88.9cm
1
Epoxy, Thermoplastic
121 lb.
23.25 in.
14.56 x 20 x 30.5 in. (37 x 50.8 x 77.5cm)
45L
77.5cm
20 in.
NFPA, OSHA
18 in.
45.7cm
Manual Close
35 in.
ChemCor™ liner, welded Steel double wall construction, 2 in. liquid tight containment sump, dual vents, adjustable shelves, three-point Stainless Steel bullet self latching doors with U-Loc™ padlockable handle, kickplat
1
Countertop storage cabinet
55kg
59.05cm
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit