Justrite™ Chemcor™ Lined Safety Cabinets for Hazardous Materials Available on GSA/VA Contract for Federal Government customers only.

Give your workers the edge. Justrite ChemCor™ thermoplastic coating on all interior surfaces provides superior protection against corrosion from drips and off-gassing from harsh chemicals.

Manufacturer: Justrite™ 8660282

 View more versions of this product

Catalog No. 19-414-812

  Check Availability
Add to Cart

    Specifications

    Color Blue
    For Use With (Application) For hazardous materials
    Capacity (Metric) 227L
    No. of Doors 2 self-closing doors
    No. of Shelves 2
    Type Lined Safety Cabinet

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit