Learn More
Justrite™ Horizontal and Vertical Drum Vents Available on GSA/VA Contract for Federal Government customers only.
For controlled release of flammable vapors; prevent explosions caused by pressure buildup
Manufacturer:  Justrite™ 08005
 View more versions of this product
Catalog No. 17-186B
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
FM
Controlled release of flammable vapors (Petroleum-based liquids), prevent explosions caused by pressure buildup
Operating valve, plated Steel flame arrester
Vertical Drum Vent
Opens at 5psig internal pressur
Vertical drums
Non-metallic Cast Iron