Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Justrite™ In-flow Safety Cans

For use in storing reagents in approved containers when drawing solvents into HPLC machines

Manufacturer:  Justrite™ 12164/DEL

 View more versions of this product

Catalog No. 19-052-390

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

  • Made of high density polyethylene
  • Equipped with a required self-closing fill/pour spout with stainless steel flame arrester
  • Two quick-disconnect fittings: one polypropylene for venting, and one stainless steel to handle the solvent in-flow
  • Stainless steel disconnect includes 1/8 in. OD (1/16 in.ID) and 1/16 in. OD (1/32 in. ID) compression fittings

Specifications

Specifications

FM, NFPA, OSHA
7.75 in.
19.6cm
24.1cm
Safety Can
5.25 in.
0.5 gal.
White
Safety
1.45 in.
3.6cm
9.5 in.
Self closing fill/pour spout
Oval
13.3cm
2L
PE (Polyethylene)
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit