Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Justrite™ QuickBerm™ Rigid Lock Spill Containment System

Catalog No. p-4844452
Click to view available options
Capacity (English):
79 gal.
119 gal.
159 gal.
175 gal.
235 gal.
318 gal.
355 gal.
398 gal.
475 gal.
745 gal.
2510 gal.
Dimensions (L x W x H):
4 x 4 x 8 ft.
4 x 6 x 8 ft.
4 x 8 x 8 ft.
8 x 8 x 8 ft.
4 x 6 x 12 ft.
4 x 8 x 12 ft.
6 x 8 x 12 ft.
8 x 10 x 8 ft.
10 x 10 x 12 ft.
12 x 28 x 12 ft.

Portable, versatile containment berm deploys quickly for emergency spill response

  • Leakproof, tough vinyl coated fabric provides broad UV and chemical resistance that withstands repeated usage
  • Reinforced inside supports reduce sagging and prevent tripping hazards that can be caused by outside supports
  • Lightweight berm foldable to small size for storage in trucks, emergency vehicles and spill kits
  • Add optional grates to help reduce contact with contaminated liquids by personnel or equipment

Applications:

  • Saddle tank ruptures
  • Personnel and small equipment decontamination
  • Fuel tank overspill protection
  • Hydraulic leaks and breaks
  • Transformer storage
  • Leaky containers, machinery, and piping
  • Overfills from aircraft or locomotive refueling
  • Truck, auto, and forklift battery storage
  • Spill kit addition

Compliance

Comply with EPA for containment and spill prevention (SPCC).

Specifications

Capacity (English) 175 gal.
Capacity (Metric) 663 L
Material PVC-coated Fabric
Type Spill Containment System
For Use With (Application) Emergency spill response
Dimensions (L x W x H) 4 x 6 x 12 ft.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.