Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Justrite™ Standard Aerosolv™ Aerosol Can Disposal System: Cartridges and Filters
For use with Serosolv Can Disposal System
Supplier: Justrite™ 28223
Description
Replaxement cartridges and filters
Specifications
Color-Changing Carbon Cartridge | |
Carbon | |
EPA, 40 CRF261.23(a)(6) | |
For Aerosolv Aerosol Can Disposal System |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction