Justrite™ Sure-Grip™ EX Hazardous Material Steel Safety Cabinet Available on GSA/VA Contract for Federal Government customers only.

Keep storage of your hazardous materials compliant with the Justrite™ Sure-Grip™ EX Hazmat Safety Cabinet.

Manufacturer: Justrite™ 866028

 View more versions of this product

Catalog No. 19-414-800

  Check Availability
Add to Cart

    Specifications

    Color Blue
    For Use With (Application) hazardous materials
    No. of Doors 2 self-closing doors
    No. of Shelves 2
    Type Safety cabinet
    Capacity 60 gal.

  • Description & Specifications
  • Promotions

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit