Learn More
JWA Mouse anti-Human, Rat, DyLight 650, Clone: S180-41, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259319C
Catalog No. NBP259319C
Add to cart
Description
Description
JWA Monoclonal antibody specifically detects JWA in Human, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
JWA
S180-41
Western Blot, Immunocytochemistry/Immunofluorescence
Purified
ARL6IP5
Fusion protein amino acids 1-524 (full length) of human EAAC1
Protein G purified
Store at 4C in the dark.
Monoclonal
Detects 60kDa.
Western Blot, Immunocytochemistry, Immunofluorescence
DyLight 650
50mM Sodium Borate with 0.05% Sodium Azide
ADP-ribosylation factor-like protein 6-interacting protein 5, ADP-ribosylation-like factor 6 interacting protein 5, Aip-5, ARL-6-interacting protein 5, Cytoskeleton-related vitamin A-responsive protein, Dermal papilla-derived protein 11, DERP11addicsin, Glutamate transporter EAAC1-interacting protein, glutamate transporter EEAC1-associated protein, GTRAP3-18aip-5, hp22, HSPC127, JM5, jmx, JWAcytoskeleton related vitamin A responsive protein, PRA1 domain family 3, PRA1 family protein 3, PRA2, PRAF3dermal papilla derived protein 11, Prenylated Rab acceptor protein 2, Protein JWa, putative MAPK activating protein PM27, Putative MAPK-activating protein PM27
Mouse
IgG1
100 ul
Primary
10550
Human, Rat