Learn More
Tocris Bioscience™ JX 06 Available on GSA/VA Contract for Federal Government customers only.
Potent and selective pyruvate dehydrogenase kinase (PDK) 1/2/3 inhibitor
$82.70 - $348.00
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
584510 Available on GSA/VA Contract for Federal Government customers only. View Documents Tocris Bioscience™
5845/10
10mg Each for $82.70
Add to cart
584550 View Documents Tocris Bioscience™
5845/50
Each for $348.00
Add to cart
Description
Description
Potent and selective pyruvate dehydrogenase kinase (PDK) 1/2/3 inhibitor (IC50 values are 49, 101 and 313 nM for PDK1, PDK2 and PDK3, respectively). Exhibits selectivity for PDK1/2/3 over PDK4 and a panel of 323 other kinases. Suppresses A549 lung cancer cell proliferation in vitro. Reduces tumor volume of A549 xenografts in mice.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Safety and Handling
Safety and Handling
Recommended Storage : Store at −20°C