Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
JX 401, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Potent, reversible inhibitor of p38α that displays no activity on the p38γ isoform (IC50 values are 32 nM and > 10 μM respectively). Inhibits the differentiation of myoblasts to myotubes in mammalian cells in culture.
Specifications
Quantity | 10 mg |
Formula Weight | 355.49 |
Percent Purity | >98% |
Chemical Name or Material | JX 401 |
Safety and Handling
Recommended Storage : Store at Room Temperature