Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JZL 195, Tocris Bioscience™

Catalog No. p-4817279
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Dual FAAH and MAGL inhibitor

Dual inhibitor of fatty acid amide hydrolase (FAAH) and monoacylglycerol lipase (MAGL) (IC50 values are 2 and 4 nM respectively). Elevates anandamide and 2-arachidonoylglycerol levels in vivo. Shown to impair short-term memory in mice.

Chemical Identifiers

CAS 1210004-12-8
Molecular Formula C24H23N3O5
Molecular Weight (g/mol) 433.464
InChI Key QNYRAEKLMNDRFY-UHFFFAOYSA-N
Synonym 4-nitrophenyl 4-3-phenoxyphenyl methyl piperazine-1-carboxylate, 4-nitrophenyl 4-3-phenoxybenzyl piperazine-1-carboxylate, d06udn, jzl hplc, 4-3-phenoxyphenyl methyl-1-piperazinecarboxylic acid 4-nitrophenyl ester
PubChem CID 46232606
IUPAC Name (4-nitrophenyl) 4-[(3-phenoxyphenyl)methyl]piperazine-1-carboxylate
SMILES C1CN(CCN1CC2=CC(=CC=C2)OC3=CC=CC=C3)C(=O)OC4=CC=C(C=C4)[N+](=O)[O-]

Specifications

Quantity 10 mg
Formula Weight 433.46
Percent Purity >98%
Chemical Name or Material JZL 195
Recommended Storage Store at -20°C
Provide Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.