Learn More
Peptides InternationalSupplier Diversity Partner JzTx-V Peptide, 5mg, MF= C157H243N47O38S7 DFS Item
Manufacturer:  Peptides InternationalSupplier Diversity Partner TOX3513PI 5MG
Jingzhaotoxin V; β/κ-theraphotoxin-Cg2a; β-theraphotoxin-Cj2a; H-Tyr-Cys-Gln-Lys-Trp-Met-Trp-Thr-Cys-Asp-Ser-Lys-Arg-Ala-Cys-Cys-Glu-Gly-Leu-Arg-Cys-Lys-Lys-Trp-Cys-Arg-Lys-Ile-Ile-OH, Application: NaV1.7 Inhibitory Peptide Toxin with IC50 Value of 0.6 nM, salt: Trifluoroacetate Salt, origin/species: Chinese earth tiger tarantula, Chilobrachys jingzhao, one letter code: H-YCQKWMWTCDSKRACCEGLRCKKWCRKII-OH, MW: 3621.43, Disulfide bonds: Cys1-Cys4, Cys2-Cys5, Cys3-Cys6
Catalog No. 50-195-2442