Learn More
Peptides InternationalSupplier Diversity Partner Tarantula Venom-Derived JzTx-V Peptide Analog, 5mg, MF= C155H236N47O41S6Br DFS Item
Manufacturer:  Peptides InternationalSupplier Diversity Partner TOX3049PI5MG
Am-6120; Glu-[Pra1; Nle6; Ala12; 5BrW24; [Glu28]JzTx-V(1-29); Tarantula Venom-Derived Nav1.7 Inhibitory JzTx-V peptide 5-Br-Trp24 Analog; JzTx-V Analog, Application: Notes: A tarantula venom-derived JzTx-V Peptide analog that blocks histamine-induced pruritis in mice, following subcutaneious administration in a NaV1.7 dependent model., salt: Trifluoroacetate Salt, origin/species: Tarantula, MW: 3686.19
Catalog No. 50-195-2440