Learn More
Active MotifSupplier Diversity Partner K-562 nuclear extract (TPA stimulated)  DFS Item
Manufacturer:  Active MotifSupplier Diversity Partner 36070
Active Motif's K-562 nuclear extract (TPA stimulated) was prepared from a cell culture of the K-562 human myelogenous leukaemia cell line. The K-562 cell line was originally derived from immortalized erythroleukaemia cells obtained from a patient with a Philadelphia chromosome-positive chronic myeloid leukemia in blast crisis. These pluripotent cells are widely applicable in research due to their ability to spontaneously differentiate into erythrocyte, granulocyte and monocyte progenitors. K-562 cells are also frequently utilized in in vitro natural killer (NK) cell assays as highly sensitive targets since they lack the expression of MHC class I molecules required for protection against NK activity. Treatment of K-562 cells with the potent tumor-promoting phorbol ester 12-O-tetradecanoylphorbol-13-acetate (TPA) leads to […] For more information, please visit: https://www.activemotif.com/catalog/details/36070.
Catalog No. NC1480842