Learn More
K'NEX Education™ Roller Coaster Physics Products for Science Education
Hands-on explorations into classic mechanics concepts.
Manufacturer:  K'Nex Education 78880
Catalog No. S90157
  Check Availability
Add to cart
Includes:
  • Teacher guide with background information and teacher notes included; aligned to state and national standards.
Specifications
Description
  • Builds 11 models, including two roller coaster designs and ramp and loop systems
  • Builds one roller coaster design or any two ramp/loop systems simultaneously
  • For grades 11 to 12
Key Concepts Include:
  • Measurement in 3-D (Trigonometry)
  • Uniform Acceleration
  • Designing Experiments
  • Projectile Motion
  • Centripetal Force and Acceleration
  • Centripetal Force in a Vertical Direction
  • Weightiness and Weightlessness
  • And much more
Description & Specifications
Specifications
Use the set to study physics and in particular mechanics
11 to 12
Teachers guide with background information and teacher notes included, aligned to state and national standards
Roller coaster physics set