Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ K252c, 99%

Inhibits protein kinase C

Manufacturer:  Alfa Aesar™ J63090MCR

 View more versions of this product

Catalog No. AAJ63090MCR


Order in Bulk

Request bulk or custom formats

Add to cart

Specifications

Specifications

1mg
-30°C to -10°C
K252c
99%
C20H13N3O
Staurosporinone; 6,7,12,13-Tetrahydro-5H-indolo[2,3-a]pyrrolo[3,4-c]carbazol-5-one
Off-white to brown
Crystalline solid
Inhibits protein kinase C
85753-43-1
311.34
Soluble in DMSO at 25mM. Also soluble in methanol. Insoluble in water.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit