Learn More
UbpbioSupplier Diversity Partner K63-UB(2-12), 50UG DFS Item
Manufacturer:  UbpbioSupplier Diversity Partner D2601
Ubiquitin/SUMO Chains – K63-Ub(1-12) (UBPBio catalog# D2600, D2601) – Enzymatically synthesized. Can be used for determination of Ub chain binding and deubiquitination assays. Ub chains are formed by conjugating the C-terminal glycine residue of Ub onto any of seven internal lysine residues or the amino group of the previous Ub. Ub chains are classified by the lysine residue used to link Ubs; different Ub chain topologies can result in different signals. For instance, Ub chains linked through lysine 6, 11, 27, 29, 33 and 48 are capable of targeting proteins for proteasomal degradation; in contrast, Ub chains linked through lysine 63 or the N-terminal amino group (linear Ub chains) often play important nonproteolytic functions including regulation of kinase activation and protein translation.
Catalog No. 50-112-6874