Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KA2/GRIK5/Glutamate Receptor KA2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Specifications
Antigen | KA2/GRIK5/Glutamate Receptor KA2 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 1:300-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP146488
|
Novus Biologicals
NBP146488 |
0.1 mL | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
NB018618
|
Novus Biologicals
NBP1464880.025ML |
0.025 mL | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
KA2/GRIK5/Glutamate Receptor KA2 Polyclonal antibody specifically detects KA2/GRIK5/Glutamate Receptor KA2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
KA2/GRIK5/Glutamate Receptor KA2 | |
Western Blot 1:300-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
Polyclonal | |
Antisera | |
RUO | |
Whole antisera with No Preservative | |
2901 | |
A synthetic peptide from 1t extracellular domain of human Glutamate Receptor KA2 (EAA2, Glutamate receptor KA-2) conjugated to an immunogenic carrier protein was used as the antigen. | |
Primary | |
Reconstitute with 0.1 ml sterilized water to desired concentration. | |
Specific for Glutamate Receptor KA2. |
LYOPH | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
EAA2, Excitatory amino acid receptor 2, glutamate receptor KA2, Glutamate receptor KA-2, glutamate receptor, ionotropic kainate 5, glutamate receptor, ionotropic, kainate 5, KA2GRIK2 | |
GRIK5 | |
IgG | |
Unpurified | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title