Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kaiso Mouse anti-Mouse, Clone: 6F1D, Active Motif™

Mouse Monoclonal Antibody

Manufacturer:  Active MotifSupplier Diversity Partner 39366

Encompass

 View more versions of this product

Catalog No. 50-199-2698


Add to cart

Description

Description

This gene encodes a transcriptional regulator with bimodal DNA-binding specificity, which binds to methylated CGCG and also to the non-methylated consensus KAISO-binding site TCCTGCNA. The protein contains an N-terminal POZ/BTB domain and 3 C-terminal zinc finger motifs. It recruits the N-CoR repressor complex to promote histone deacetylation and the formation of repressive chromatin structures in target gene promoters. It may contribute to the repression of target genes of the Wnt signaling pathway, and may also activate transcription of a subset of target genes by the recruitment of catenin delta-2 (CTNND2). Its interaction with catenin delta-1 (CTNND1) inhibits binding to both methylated and non-methylated DNA. It also interacts directly with the nuclear import receptor Importin-alpha 2 (also known as karyopherin alpha2 or RAG cohort 1), which may mediate nuclear import of this protein. Alternatively spliced transcript variants encoding the same protein have been identified.
Specifications

Specifications

Kaiso
Western Blot
1 mg/mL
PBS with 30% glycerol and 0.035% sodium azide; pH 7.5
Q8BN78
AW260253; E130014G12Rik; KAISO; kaiso transcription factor; RGD1566309; RP23-322E15.9; Transcriptional regulator Kaiso; WUGSC:H_DJ525N14.1; ZBTB33; zinc finger and BTB domain containing 33; Zinc finger and BTB domain-containing protein 33; zinc finger transcription factor Kaiso; ZNF348; Znf-kaiso
This kaiso antibody was raised against a His-Tagged fusion protein corresponding to amino acids 103-499 of mouse kaiso.
Protein A
RUO
Antibody
Monoclonal
Mouse
ZBTB33
6F1D
Unconjugated
ZBTB33
Liquid
Mouse
IgG
10 μg
-20°C, Avoid Freeze/Thaw Cycles
Primary
56805
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit