Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Kallikrein 13 Antibody Pair [HRP]

Polyclonal/Monoclonal Antibody Pair

Manufacturer:  Novus Biologicals™ NBP2795095PLATES

 View more versions of this product

Catalog No. NB010058


Add to cart

Description

Description

This solid phase sandwich ELISA utilizes an antibody specific for Human Kallikrein 13.
Specifications

Specifications

HRP
Antibody Pairs
39.0625 pg/mL
26085
RUO
Kallikrein 13
Cancer
Human
KLK13
1. Capture Antibody: Dilute to a working concentration of 2.0 μg/mL in CBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute standard powder with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve usi ng 2-fold serial dilutions in sample dilution buffer, and a high standard of 2500 pg/mL is recommended.
Sandwich ELISA
This solid phase sandwich ELISA utilizes an antibody specific for Human Kallikrein 13.
Sandwich ELISA
  • Detection Antibody: 0.5 mg/mL rabbit anti-Kallikrein 13 polyclonal antibody conjugated to horseradish-peroxidase (HRP).
  • Capture Antibody: 0.25 mg/mL of mouse anti-Kallikrein 13 monoclonal antibody.
  • Standard: Each vial contains 110 ng of recombinant Kallikrein 13.
  • Storage is content dependent.
    Rabbit/Mouse
    5 Plates
    39.06-2500 pg/mL
    DKFZp586J1923, EC 3.4.21, EC 3.4.21.-, kallikrein 13, Kallikrein-like protein 4, kallikrein-related peptidase 13, KLK-L4kallikrein-13, KLKL4kallikrein-like gene 4
    SDS
    Documents
    Product Certifications
    Promotions

    Promotions

    Provide Content Correction

    We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Cancel Submit