Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Kallikrein 13 Antibody Pair [HRP]
Polyclonal/Monoclonal Antibody Pair
$1183.00
Specifications
Assay Range | 39.0625 pg/mL |
---|---|
Assay Sensitivity | 39.06-2500 pg/mL |
Assay | Sandwich ELISA |
Conjugate | HRP |
Product Type | Antibody Pairs |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB010057
|
Novus Biologicals™
NBP27950915PLATES |
15 Plates |
Each of 1 for $1,183.00
|
|
|||||
Description
This solid phase sandwich ELISA utilizes an antibody specific for Human Kallikrein 13.Specifications
39.0625 pg/mL | |
Sandwich ELISA | |
Antibody Pairs | |
Human | |
DKFZp586J1923, EC 3.4.21, EC 3.4.21.-, kallikrein 13, Kallikrein-like protein 4, kallikrein-related peptidase 13, KLK-L4kallikrein-13, KLKL4kallikrein-like gene 4 | |
KLK13 | |
Cancer | |
Kallikrein 13 |
39.06-2500 pg/mL | |
HRP | |
This solid phase sandwich ELISA utilizes an antibody specific for Human Kallikrein 13. | |
Sandwich ELISA | |
26085 | |
Rabbit/Mouse | |
RUO | |
Storage is content dependent. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title