Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kallikrein 14 Goat anti-Human, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Supplier: R&D Systems AF2626SP
This item is not returnable.
View return policy
Description
Kallikrein 14 Polyclonal specifically detects Kallikrein 14 in Human samples. It is validated for Western Blot, Immunoprecipitation, Neutralization, ELISA Capture (Matched Antibody Pair).Specifications
Kallikrein 14 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
EC 3.4.21, EC 3.4.21.-, hK14, kallikrein 14, Kallikrein-like protein 6, kallikrein-related peptidase 14, KLK14, KLKL6, KLK-L6kallikrein-14 | |
Goat | |
Antigen Affinity-purified | |
RUO | |
43847 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunoprecipitation, Neutralization, ELISA | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL, Neutralization 1 ug/mL, ELISA Capture (Matched Antibody Pair) 0.2-0.8 ug/mL | |
AAD50773 | |
KLK14 | |
Mouse myeloma cell line NS0-derived recombinant human Kallikrein 14 Gln19-Met248, predicted Accession # AAD50773 | |
25 μg | |
Primary | |
Detects human Kallikrein 14 in ELISAs and Western blots. | |
Human | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only