Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Kallikrein 14 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA14240
Description
PA1-4240 detects Kallikrein-14 from human samples. PA1-4240 has been successfully used in Western blot procedures. PA1-4240 immunizing peptide corresponds to amino acid residues 239-251from human Kallikrein-14.
Kallikreins are a subgroup of serine proteases and are implicated in carcinogenesis. A novel KLK gene, KLK12, is expressed in a variety of tissues including salivary gland, stomach, uterus, lung, thymus, prostate, colon, brain, thyroid, and trachea. It has applications as a novel cancer biomarker.
Specifications
Kallikrein 14 | |
Polyclonal | |
Unconjugated | |
Klk14 | |
GK14; Glandular kallikrein KLK14; hK14; kallikrein 14; Kallikrein L6; kallikrein related peptidase 14; kallikrein related-peptidase 14; Kallikrein-14; kallikrein-like protein 6; kallikrein-related peptidase 14; KLK14; KLK-14; KLKL6; KLK-L6; mGK14 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
43847 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q9P0G3 | |
Klk14 | |
Synthetic Peptide corresponding to residues K(239) Y R S W I E E T M R D K(251) of human Kallikrein-14. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction