Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kallikrein 14 Rabbit anti-Human, Alexa Fluor 647, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB200132AF647
Description
Kallikrein 14 Polyclonal antibody specifically detects Kallikrein 14 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)Specifications
Kallikrein 14 | |
Polyclonal | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
Alexa Fluor 647 | |
EC 3.4.21, EC 3.4.21.-, hK14, kallikrein 14, Kallikrein-like protein 6, kallikrein-related peptidase 14, KLKL6, KLK-L6kallikrein-14 | |
Residues 239-251 [KYRSWIEETMRDK] of the human KLK14 protein | |
0.1 mL | |
Cancer | |
43847 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction