Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kallikrein 3/PSA Rabbit anti-Human, Clone: 103, Novus Biologicals™
SDP

Rabbit Monoclonal Antibody

$340.00 - $521.00

Specifications

Antigen Kallikrein 3/PSA
Clone 103
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin 1:100-1:500
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB020750
SDP
View Documents
Novus Biologicals
NBP28963350UL
50 μg
Each for $340.00
Add to Cart
 
NB020749
SDP
View Documents
Novus Biologicals
NBP289633100UL
100 μg
Each for $521.00
Add to Cart
 
Description

Description

Kallikrein 3/PSA Monoclonal specifically detects Kallikrein 3/PSA in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

Kallikrein 3/PSA
Immunohistochemistry, Immunohistochemistry-Paraffin 1:100-1:500
Monoclonal
Purified
RUO
Human
APS seminin, EC 3.4.21, EC 3.4.21.77, Gamma-seminoprotein, hK3, kallikrein 3, (prostate specific antigen), Kallikrein-3, kallikrein-related peptidase 3, KLK2A1, P-30 antigen, prostate specific antigen, prostate-specific antigen, PSA, PSA semenogelase, Semenogelase, Seminin
Recombinant Human Kallikrein 3/PSA protein
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
103
Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Cancer, Cellular Markers, Prostate Cancer, Tumor Biomarkers
0.2 um filtered solution in PBS
354
IgG
Protein A purified
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit