Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Kallikrein 4 Monoclonal Antibody (16)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA530963
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human KLK4/Kallikrein 4.
Kallikreins are a subgroup of serine proteases and are implicated in carcinogenesis. A novel Kallikrein gene Prostase/KLK-L1 (also known as KLK4) is expressed and is up-regulated by androgens and progestins. KLK-L1 may be involved in the pathogenesis and/or progression of prostate, breast, and possibly other malignancies.
Specifications
Kallikrein 4 | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
Q9Y5K2 | |
KLK4 | |
Recombinant Human KLK4/Kallikrein 4 Protein (Met1-Ser254). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2b |
ELISA | |
16 | |
Unconjugated | |
KLK4 | |
AI2A1; androgen-regulated message 1; ARM1; EMSP; Emsp1; enamel matrix serine protease 1; Enamel matrix serine proteinase 1; ESMP1; kallikrein; kallikrein 4 (prostase, enamel matrix, prostate); kallikrein related peptidase 4; kallikrein related-peptidase 4 (prostase, enamel matrix, prostate); kallikrein-4; Kallikrein-like protein 1; kallikrein-related peptidase 4; Klk4; KLK-4; KLKL1; KLK-L1; Prostase; protease, serine 17 (enamel matrix, prostate); Prss17; PSTS; serine protease 17 | |
Mouse | |
Protein A | |
RUO | |
9622 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction