Learn More
Kallikrein 4/Prostase/EMSP1 Mouse anti-Human, Alexa Fluor 700, Clone: MM0431-5D34, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP211751AF700
Catalog No. NB211751700
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Kallikrein 4/Prostase/EMSP1 Monoclonal antibody specifically detects Kallikrein 4/Prostase/EMSP1 in Human samples. It is validated for Western Blot.
Specifications
Specifications
Kallikrein 4/Prostase/EMSP1
MM0431-5D34
Western Blot
Purified
KLK4
Human Kallikrein-4
Protein G purified
Apoptosis, Cancer, Tumor Suppressors
Primary
9622
Western Blot
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
AI2A1, ARM1, EC 3.4.21, EC 3.4.21.-, EMSP, EMSP1MGC116827, Enamel matrix serine proteinase 1, kallikrein 4 (prostase, enamel matrix, prostate), kallikrein-4, Kallikrein-like protein 1, kallikrein-related peptidase 4, KLK-L1MGC116828, Prostase, PRSS17enamel matrix serine protease 1, PSTSandrogen-regulated message 1, Serine protease 17
Mouse
IgG2
0.1 ml
Store at 4C in the dark.
Monoclonal
Human