Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kallikrein 5 Mouse anti-Human, Clone: KLK5/3843, Novus Biologicals™

Mouse Monoclonal Antibody

$138.37 - $352.89

Specifications

Antigen Kallikrein 5
Clone KLK5/3843
Concentration 0.2 mg/ml
Dilution Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array
Applications Immunohistochemistry (Paraffin), Peptide Array
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB123192
View Documents
Novus Biologicals
NBP30785920UG
20 μg
Each for $138.37
Add to cart
 
NB123191
View Documents
Novus Biologicals
NBP30785910UG
100 μg
Each for $352.89
Add to cart
 
Description

Description

Kallikrein 5 Monoclonal antibody specifically detects Kallikrein 5 in Human samples. It is validated for Immunohistochemistry (Paraffin), Protein Array
Specifications

Specifications

Kallikrein 5
0.2 mg/ml
Immunohistochemistry (Paraffin), Peptide Array
Purified
RUO
Human
EC 3.4.21, EC 3.4.21.4, kallikrein 5, Kallikrein-like protein 2, kallikrein-related peptidase 5, KLKL2, KLK-L2EC 3.4.21.-, SCTEkallikrein-5, Stratum corneum tryptic enzyme
Recombinant fragment of human Kallikrein 5 protein (around aa 36-177) (exact sequence is proprietary)
Monoclonal
Protein A or G purified
KLK5/3843
Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array
Unconjugated
Mouse
Cancer
10 mM PBS with 0.05% BSA
25818
IgG2b Kappa
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit