Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kallikrein 8/Neuropsin Antibody (140), PE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289988PE
Description
Kallikrein 8/Neuropsin Monoclonal antibody specifically detects Kallikrein 8/Neuropsin in Human samples. It is validated for Western BlotSpecifications
Kallikrein 8/Neuropsin | |
Monoclonal | |
PE | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot | |
140 | |
Western Blot | |
EC 3.4.21, EC 3.4.21.118, hK8, HNP, kallikrein 8 (neuropsin/ovasin), kallikrein-related peptidase 8, KLK8, KLK8 protein type 1, KLK8 protein type 2, Neuropsin, neuropsin type 1, neuropsin type 2, NP, NRPN, Ovasin, PRSS19kallikrein-8, Serine protease 19, Serine protease TADG-14, TADG14neuropsin, Tumor-associated differentially expressed gene 14 protein | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human Kallikrein 8/Neuropsin (Uniprot#: O60259-1; Met1-Gly260). | |
0.1 mL | |
Neuroscience | |
11202 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction