Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Kanamycin Sulfate, Ultrapure, Thermo Scientific™

Click to view available options
Quantity:
5g
25g
Packaging:
Plastic bottle
25389-94-0
C18H38N4O15S
582.575
MFCD00070253
OOYGSFOGFJDDHP-IZQIRFRQSA-N
Kanamycin A
134129479
(2R,3S,4S,5S,6R)-2-(aminomethyl)-6-[(1R,2S,3S,4R,6S)-4,6-diamino-3-[(2S,3R,4S,5R,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-2-hydroxycyclohexyl]oxyoxane-3,4,5-triol;sulfuric acid
C1C(C(C(C(C1N)OC2C(C(C(C(O2)CN)O)O)O)O)OC3C(C(C(C(O3)CO)O)N)O)N.OS(=O)(=O)O
25389-94-0
C18H38N4O15S
582.575
MFCD00070253
OOYGSFOGFJDDHP-IZQIRFRQSA-N
Kanamycin A
134129479
(2R,3S,4S,5S,6R)-2-(aminomethyl)-6-[(1R,2S,3S,4R,6S)-4,6-diamino-3-[(2S,3R,4S,5R,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-2-hydroxycyclohexyl]oxyoxane-3,4,5-triol;sulfuric acid
C1C(C(C(C(C1N)OC2C(C(C(C(O2)CN)O)O)O)O)OC3C(C(C(C(O3)CO)O)N)O)N.OS(=O)(=O)O
  • Selectable marker for cosmid vectors used in transfection of mammalian cells
  • Mode of Action: Binds to ribosomal components and inhibits protein synthesis
  • Kanamycin is inactivated by aminoglycoside phospho transferase (MW 25,000) in
Packaging Plastic bottle
Formula Weight 582.58
Physical Form Powder
Quantity 25g
Chemical Name or Material Kanamycin sulfate