Learn More
KANK1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 205471AP
Catalog No. 50-173-2952
Add to cart
Description
Description
The antibody is specific to KANK1.
Ankyrins are membrane adaptor molecules that play important roles in the control of cytoskeleton formation by regulating actin polymerization. KANK1 (KN motif and ankyrin repeat domain-containing protein 1), also known as ANKRD15, is a 1,352 amino acid protein that contains at least 12 exons and 5 ANK repeats. It binds to beta-catenin and regulates its subcellular distribution. KANK1 is ubiquitously expressed and localizes to cytoplasm. It may function as a tumor suppressor for renal cell carcinoma. Mutations in this gene cause cerebral palsy spastic quadriplegic type 2, a central nervous system development disorder.
Specifications
Specifications
KANK1
0.19 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q14678
ANKRD15, KANK, KANK1, KIAA0172
Rabbit
IgG
150 μL
-20°C
Primary
107351, 23189, 309429
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
KANK1
Liquid
Kank1
Peptide
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat