Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kaolin, USP, Fisher Chemical

Catalog Number k2500
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
1332-58-7
Al2H4O9Si2
258.16
MFCD00062311
NLYAJNPCOHFWQQ-UHFFFAOYSA-N
kaolin, china clay, argilla, glomax, porcelain clay, bolus alba, white bole, hydrated aluminum silicate, kaolin clay slurry, alphagloss
56841936
dioxo-2,4,6-trioxa-3,5-disila-1,7-dialuminaheptane-3,5-dione dihydrate
O.O.O=[Al]O[Si](=O)O[Si](=O)O[Al]=O
1332-58-7
Al2H4O9Si2
258.16
MFCD00062311
NLYAJNPCOHFWQQ-UHFFFAOYSA-N
kaolin, china clay, argilla, glomax, porcelain clay, bolus alba, white bole, hydrated aluminum silicate, kaolin clay slurry, alphagloss
56841936
dioxo-2,4,6-trioxa-3,5-disila-1,7-dialuminaheptane-3,5-dione dihydrate
O.O.O=[Al]O[Si](=O)O[Si](=O)O[Al]=O
  • Kaolin is a powdery clay that is useful in a variety of applications due to its emulsifying effects and other material properties
  • This grade of kaolin is tested for the absence of E. coliand meets USP specifications
TRUSTED_SUSTAINABILITY
Melting Point 1760°C
Color Yellow
Physical Form Solid
Quantity 500 g
Solubility Information 2.0% max. (USP) Acid Soluble Substances
Formula Weight 258.2
Grade USP
Identification Pass Test (USP)
Loss on Ignition 15.0% max. (USP)
Packaging Poly Bottle
Density 1.8g/mL
Chemical Name or Material Kaolin
Show More Show Less
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit