Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kaolin, pure, ACROS Organics™

Click to view available options
Quantity:
1kg
2.5kg
10kg
Packaging:
Plastic Bottle
Plastic drum
1332-58-7
H2Al2Si2O8·H2O
258.2
NLYAJNPCOHFWQQ-UHFFFAOYSA-N
kaolin,china clay,argilla,glomax,porcelain clay,bolus alba,white bole,hydrated aluminum silicate,kaolin clay slurry,alphagloss
56841936
oxo-oxoalumanyloxy-[oxo(oxoalumanyloxy)silyl]oxysilane;dihydrate
O.O.O=[Al]O[Si](=O)O[Si](=O)O[Al]=O
1332-58-7
H2Al2Si2O8·H2O
258.2
NLYAJNPCOHFWQQ-UHFFFAOYSA-N
kaolin,china clay,argilla,glomax,porcelain clay,bolus alba,white bole,hydrated aluminum silicate,kaolin clay slurry,alphagloss
56841936
oxo-oxoalumanyloxy-[oxo(oxoalumanyloxy)silyl]oxysilane;dihydrate
O.O.O=[Al]O[Si](=O)O[Si](=O)O[Al]=O
Merck Index 14,5282
Solubility Information Solubility in water: insoluble. Other solubilities: insoluble in cold acids and alkali hydroxides
Formula Weight 258.2
Physical Form Powder
Grade Pure
Loss on Ignition 10.8% to 11.8%
Loss on Drying 1.5% max.
Packaging Plastic Bottle
Particle Size 1.0 to 1.8 Micron
Color Beige
Melting Point 1750.0°C
Quantity 1kg
Chemical Name or Material Kaolin
Show More Show Less