Learn More
KAP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 152021AP
Catalog No. 50-173-0062
Add to cart
Description
Description
TIF1 beta is a member of the tripartite motif (TRIM) family. It is composed of a N-terminal RBCC (RING finger, B boxes, coiled coil) motif, and a C-terminal bromodomain which is preceded by a PHD finger. TIF1 beta has been reported to function as a co-repressor for Kruppel-associated box (KRAB)-domain-containing zinc finger proteins. As such, it plays a crucial role in early embryogenesis. It appears that the co-repressor function of TIF1 beta is related to its organization and/or maintece of higher-order, heterochromatin-like structures.
Specifications
Specifications
KAP1
0.4 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O08629, Q13263, Q62318
KAP 1, KAP1, KRAB associated protein 1, KRAB interacting protein 1, KRIP 1, Nuclear corepressor KAP 1, RING finger protein 96, RNF96, TF1B, TIF1 beta, TIF1B, TRIM28, tripartite motif containing 28
Rabbit
IgG
150 μL
-20°C
Primary
10155, 116698, 21849
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation
Unconjugated
TRIM28
Liquid
TRIM28
KAP1 Fusion Protein Ag7350
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat