Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kappa Light Chain Antibody (HP6053 + L1C1), Novus Biologicals™
Mouse Monoclonal Antibody
$265.00 - $506.50
Specifications
Antigen | Kappa Light Chain |
---|---|
Clone | HP6053 + L1C1 |
Dilution | Western Blot 0.5-1.0ug/ml, Flow Cytometry 0.5-2.0ug/million cells, Immunoprecipitation 1-2ug/500ug protein lysate, Immunohistochemistry-Paraffin 0.5-1ug/ml, Immunohistochemistry-Frozen 0.5-1ug/ml |
Applications | Western Blot, Flow Cytometry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP234258F
|
Novus Biologicals
NBP2342580.02MG |
0.02 mg |
Each for $265.00
|
|
|||||
NBP234258A
|
Novus Biologicals
NBP2342580.1MG |
0.1 mg |
Each for $506.50
|
|
|||||
NBP234258B
|
Novus Biologicals
NBP2342580.2MG |
0.2 mg | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Description
Kappa Light Chain Monoclonal specifically detects Kappa Light Chain in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Kappa Light Chain | |
Western Blot 0.5-1.0ug/ml, Flow Cytometry 0.5-2.0ug/million cells, Immunoprecipitation 1-2ug/500ug protein lysate, Immunohistochemistry-Paraffin 0.5-1ug/ml, Immunohistochemistry-Frozen 0.5-1ug/ml | |
Monoclonal | |
Purified | |
RUO | |
Human | |
P01601 | |
3514 | |
Purified human Ig kappa chain (HP6053) & Human B-Lymphoma Cells (L1C1) | |
Primary | |
Store at 4C. | |
22.5 kDa |
HP6053 + L1C1 | |
Western Blot, Flow Cytometry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
Unconjugated | |
Mouse | |
Adaptive Immunity, Immunology | |
PBS with 0.05% BSA. with 0.05% Sodium Azide | |
HCAK1, IGKCD, immunoglobulin kappa constant, Km, MGC111575, MGC62011, MGC72072, MGC88770, MGC88771, MGC88809 | |
IGKC | |
IgG | |
Protein A purified | |
This MAb is specific to kappa light chain of immunoglobulin and shows no cross-reaction with lambda light chain or any of the five heavy chains. In mammals, the two light chains in an antibody are always identical, with only one type of light chain, kappa or lambda. The ratio of Kappa to Lambda is 70:30. However, with the occurrence of multiple myeloma or other B-cell malignancies this ratio is disturbed. Antibody to the kappa light chain is reportedly useful in the identification of leukemias, plasmacytomas, and certain non-Hodgkin's lymphomas. Demonstration of clonality in lymphoid infiltrates indicates that the infiltrate is malignant. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title