Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kappa Light Chain Mouse anti-Human, PE/Atto594, Clone: KLC1278, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP254512PEATT594

Catalog No. N254512P594


Add to cart

Description

Description

Kappa Light Chain Monoclonal antibody specifically detects Kappa Light Chain in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

Kappa Light Chain
KLC1278
Flow Cytometry
Conjugated, Purified
IGKC
Recombinant human full-length Ig kappa chain
Protein A or G purified
Adaptive Immunity, Immunology
Primary
3514.0
Flow Cytometry
PE/ATTO 594
PBS with 0.05% Sodium Azide
HCAK1, IGKCD, immunoglobulin kappa constant, Km, MGC111575, MGC62011, MGC72072, MGC88770, MGC88771, MGC88809
Mouse
IgG1 Kappa
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit