Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kappa Opioid Receptor Rabbit anti-Rat, Polyclonal, Neuromics™ Non-distribution product as customer accommodation.

Rabbit Polyclonal Antibody

$166.40 - $419.12

Specifications

Antigen Kappa Opioid Receptor
Concentration Conc. Not Determined
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-210-0022 Non-distribution product as customer accommodation.
View Documents
Neuromics, IncSupplier Diversity Partner
RA10103T150UL
150 μL
Each for $419.12
Add to cart
 
50-210-0023 Non-distribution product as customer accommodation.
View Documents
Neuromics, IncSupplier Diversity Partner
RA10103T50UL
50 μL
Each for $166.40
Add to cart
 
Description

Description

Kappa Opioid Receptor Polyclonal antibody specifically detects Kappa Opioid Receptor in Rat samples. It is validated for Immunocytochemistry, Immunofluorescence
Specifications

Specifications

Kappa Opioid Receptor
Immunocytochemistry, Immunofluorescence
Unconjugated
Rabbit
Rat
P34975
29335
IgG
Primary
Antibody
Conc. Not Determined
Polyclonal
Liquid
RUO
whole serum with 0.05% sodium azide
kappa opioid receptor; Kappa-type opioid receptor; KOPR1; KOR; K-OR-1; KOR-1; MSL-1; Opiate receptor, kappa-1; opioid receptor kappa 1; opioid receptor, kappa 1; opioid receptor, kappa 2; OPRK; Oprk1; Oprk2; Ork1; R21; Ror-d
DPAYLRDIDGMNKPV-residues 366-380 of the human Kappa Opioid Receptor.
Polyclonal
-20° C, Avoid Freeze/Thaw Cycles
OPRK1
SDS
Product Certifications
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit