Kappler Frontline 300 Chemical and Fire Protection Coverall with Hood Opening

Manufacturer: Kappler, Inc.Supplier Diversity Partner F3H429 SI 4X

 View more versions of this product

Catalog No. 18-025-674

  Check Availability
Add to Cart

    Specifications

    Certifications/Compliance ASTM F1001
    Material Frontline™ 300
    Includes Front entry, expanded back, double-taped seams, 2 exhalation valves, attached sock boots with boot flap, Butyl gloves with specially attached barrier glove liner and knitted Kevlar™ overglove, Non-skid sole, cone Inserts
    Seam Type Bound, piped, sealed
    Color Silver
    Size 4X-Large
    For Use With (Application) Ideal for petrochemical operations, line maintenance, tank cleaning, refueling situations
    Resistance Fire, chemical
    Closure Type Double Storm Flap with Hook and Loop

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit