Kappler Frontline™ 500 Vapor Total Encapsulating Suit
Broad chemical holdout plus flame resistance and radiant heat protection for additional hazards encountered in a chemical flash-fire
Manufacturer: Kappler, Inc.Supplier Diversity Partner F5H580 91 SI 4X
 View more versions of this product
Catalog No. 18-025-693
  Check Availability
Add to Cart
    Specifications
    Certifications/Compliance NFPA 1991
    Material Frontline™ 500 FR
    Includes Front entry, expanded back, double-taped seams, 2 exhalation valves, double storm flap with hook and loop closure, attached sock boots with boot flap,butyl gloves with specially attached barrier glove liner and knitted Kevlar™ overglove
    Seam Type Heat sealed, taped
    Color Silver
    Size 4X-Large
    For Use With (Application) Petrochemical operations, line maintenance, tank cleaning, refueling situations, hazmat response, chemical handling with potential for chemical flash-fire
    Closure Type Hook and loop
  • Description & Specifications
Related Products
Product Content Feedback