Learn More
Kappler Frontline™ 500 Vapor Total Encapsulating Suit
Broad chemical holdout plus flame resistance and radiant heat protection for additional hazards encountered in a chemical flash-fire
Manufacturer:  Kappler, Inc.Supplier Diversity Partner F5H580 91 SI SMMD
 View more versions of this product
Catalog No. 18-025-690
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
NFPA 1991
Frontline™ 500 FR
Heat sealed, taped
Small, Medium
Hook and loop
Front entry, expanded back, double-taped seams, 2 exhalation valves, double storm flap with hook and loop closure, attached sock boots with boot flap,butyl gloves with specially attached barrier glove liner and knitted Kevlar™ overglove
Silver
Petrochemical operations, line maintenance, tank cleaning, refueling situations, hazmat response, chemical handling with potential for chemical flash-fire