Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kappler ProVent 10,000 Limited-Use Lab Coats
Comfortable fit in a tailored style
$414.00 - $476.00
Specifications
Certifications/Compliance | ASTM 1670, ASTM 1671 |
---|---|
Closure Type | Snaps |
Material | ProVent™ fabric |
No. of Pockets | 3 |
Disposable | Yes |
Catalog Number | Mfr. No. | Color | Size | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Size | Price | Quantity & Availability | ||||
19-315349
|
Kappler
PPS235 BU 2X |
Blue | 2X-Large |
Case of 30 for $455.00
|
|
19-315346
|
Kappler
PPS235 BU MD |
Blue | Medium |
Case of 30 for $414.00
|
|
19-315348
|
Kappler
PPS235 BU XL |
Blue | X-Large |
Case of 30 for $414.00
|
|
19-315345
|
Kappler
PPS235 BU SM |
Blue | Small |
Case of 30 for $414.00
|
|
19-315347
|
Kappler
PPS235 BU LG |
Blue | Large |
Case of 30 for $414.00
|
|
17-988-491A
|
Kappler
PPS235WHSM |
White | Small |
Case of 30 for $414.00
|
|
17-988-491B
|
Kappler
PPS235WHMD |
White | Medium |
Case of 30 for $414.00
|
|
17-988-491C
|
Kappler
PPS235WHLG |
White | Large |
Case of 30 for $414.00
|
|
17-988-491D
|
Kappler
PPS235WHXL |
White | X-Large |
Case of 30 for $414.00
|
|
17-988-491F
|
Kappler
PPS235WH3X |
White | 3X-Large |
Case of 30 for $476.00
|
|
17-988-491E
|
Kappler
PPS235WH2X |
White | 2X-Large |
Case of 30 for $455.00
|
|
19-315350
|
Kappler
PPS235 BU 3X |
Blue | 3X-Large |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
17-988-491G
|
Kappler
PPS235 WH 4X |
White | 4X-Large |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
Description
Patented microporous film composite fabric offers a wide range of barrier protection against blood, body fluids and viral threats.
- High moisture vapor transmission rate provides cool, comfort
- Ideal for biohazard cleanup
- Six snaps for front closure
- Serged-seam stitching, breast pocket, two hip pockets
- Knit collar and cuffs
Compliance
Passes ASTM 1670 blood penetration and ASTM 1671 viral penetration tests
Specifications
ASTM 1670, ASTM 1671 | |
ProVent™ fabric | |
Yes | |
Non-sterile | |
8.2 kg |
Snaps | |
3 | |
Serged | |
18 lb. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title