Learn More
Kappler™ ProVent™ 10000 Gowns
Wrap-Around Gown, Hook and Loop Neck Closure, Two Waist Ties, Knit Cuffs, Non-Sterile
Manufacturer: Kappler™ MN101BLUE3X
This item is not returnable.
View return policy
Description
- ProVent 10,000 is a patented microporous film composite fabric that offers a wide range of barrier protection against blood-borne pathogens, body fluids and viral threats.
- It passes both ASTM F1670 blood penetration and ASTM F1671 viral penetration tests.
- The high moisture vapor transmission rate (MVTR) of ProVent 10,000 results in a more comfortable garment ideal for biohazard work applications.
Compliance
Meets ASTM™ F1670 and ASTM F1671 standards.
Specifications
3X-Large | |
Blue | |
Non-sterile | |
Biohazard cleanup | |
Ultrasonic |
Composite Fabric | |
Wrap around | |
Wrap-around | |
Knit | |
ASTM F1670, ASTM F1671 |