Kappler Provent 10000 Open-Back Gowns

Manufacturer: Kappler, Inc.Supplier Diversity Partner MN108BUMD

 View more versions of this product

Catalog No. 17-988-496B

  Check Availability
Add to Cart

    Specifications

    For Use With (Application) Used in general purpose
    Size Medium
    Seam Type Ultrasonic
    Sterile No
    Weight (English) 16 lb.
    Weight (Metric) 7.2kg
    Cuff Style Knit
    Certifications/Compliance ASTM 1670, ASTM 1671
    Closure Type Hook and loop
    Color Blue
    Material Composite Fabric

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit