Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kappler™ ProVent™ 7000 Isolation Gowns

Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Weight (English):
16 lb.
16 lbs.
17 lb.
17 lbs.
Cuff Style:
Elastic
Knit
This item is not returnable. View return policy
This item is not returnable. View return policy

Resistant to abrasions, punctures, and tears

Lightweight composite fabric utilizes microporous film technology to provide excellent protection against hazardous particles and light liquid splash and maintain strength wet or dry.

  • Wraparound style opens to back
  • Knit cuffs or elastic wrists
  • Two neck ties, two waist ties
  • Nonsterile
  • White

Order Info

  • Front length: small 38.75 in. (98.4 cm), medium 39.75 in. (101 cm), large 40.75 in. (103.5 cm). Back length of all sizes: 3 in. (7.6 cm) longer than front

Compliance

Meets ASTM™ F1670 and ASTM F1671 standards.

Size Small
Material Composite Fabric
Color White
Sterility Non-sterile
Style Wrap-around
For Use With (Application) Protection from aerosols and liquids in clinical settings
Cuff Style Knit
Seam Type Serged
Certifications/Compliance ASTM F1670, ASTM F1671
Weight (English) 17 lbs.