Learn More
Kappler™ Zytron™ 100 Chemical-Resistant Coveralls
Provides basic chemical resistance
Manufacturer:  Kappler™ Z1B4173X
 View more versions of this product
Catalog No. 19-167-838F
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Zippered front
  • Seam type available in bound and serged
  • Available with attached hood and/or boots
  • Color: Yellow
Ideal for applications where workers need protection from dry particulates and light splash
Compliance
Meets ASTM™ F739 industrial chemical testing requirements.
Specifications
Specifications
ASTM F739
Bound
3X-Large
11 lb.
Zipper front
Collar, elastic wrists and ankles
Elastic
Yellow
Ideal for applications where workers need protection from dry particulates and light splash
5kg
Elastic
Zytron™ 100