Learn More
Kappler Zytron™ 200 Hooded Coveralls
Provide increased protection with resistance to a wider range of chemicals
Manufacturer:  Kappler, Inc.Supplier Diversity Partner Z2H419 GY LGXL
 View more versions of this product
Catalog No. 19-167-811F
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Double Storm Flap with Hook and Loop
Zytron™ 200
Heat sealed, taped
Large, X-Large
Chemical
Multilayer film barrier laminated to a 1.5 oz./yd2 Polypropylene substrate
Attached hood and boots, elastic wrist, double storm flap with hook and loop closure over the front zipper
Gray
Ideally used with wider range of chemicals