Learn More
Kappler™ Zytron™ 200 Hooded Coveralls
Manufacturer:  Kappler™ Z2H419 GY 2X3X
Catalog No. 19-167-811G
Add to cart
This item is not returnable. View return policy
Description
Description
Specifications
Specifications
Zipper with hook-and-loop seal and double storm flap
Zytron™ 200
Gray
Attached hood and boots, elastic wrist, double storm flap with hook and loop closure over the front zipper; Multilayer film barrier laminated to a 1.5 oz./yd2 Polypropylene substrate
Hooded
Sealed, Taped
Attached
Attached boots
Elastic
Ideally used with wider range of chemicals
Chemical
2X-Large, 3X-Large