Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

Kappler™ Zytron™ 200 Hooded Coveralls

Provide increased protection with resistance to a wider range of chemicals

Manufacturer:  Kappler™ Z2H414XL

 View more versions of this product

Catalog No. 19-171-140C



Description

Description

  • Attached hood
  • Zippered front
  • Color: Gray

Applications:

Valuable in applications where rip-out is a concern

Specifications

Specifications

ASTM D751, D3787
Heat-sealed and taped seams, Taped
X-Large
Ideally used with wider range of chemicals
4.1kg
Attached boots
Zipper front, attached hood and boots, elastic wrist and face opening, double storm flap with hook and loop closure
Elastic
Gray
Elastic
9 lb.
Zipper, Double storm flap with hook and loop
Hooded
Zytron™ 200
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit