Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kappler™ Zytron™ 200 Hooded Coveralls

Provide increased protection with resistance to a wider range of chemicals

Manufacturer:  Kappler™ Z2H4283X

 View more versions of this product

Catalog No. 19-171-143E


Add to cart

Description

Description

  • Attached hood
  • Zippered front
  • Color: Gray

Applications:

Valuable in applications where rip-out is a concern

Specifications

Specifications

ASTM D751, D3787, CE Type 3
Heat-sealed and taped seams, Taped
3X-Large
Ideally used with wider range of chemicals
4.1kg
Elastic
Elastic ankle and cuff, double storm flap with hook and loop closure over the front zipper, attached hood, face opening
Elastic
Gray
Hooded
9 lb.
Double storm flap with hook and loop
Hooded
Zytron™ 200
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit